Anti-CD9 Mouse Monoclonal Antibody [clone: 4A2]

Supplier: Abnova
H00000928-M01
10581-546EA 335 USD
10581-546
Anti-CD9 Mouse Monoclonal Antibody [clone: 4A2]
Antibodies
Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: 4A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR