Anti-CD8B Rabbit Polyclonal Antibody

Supplier: Abnova
H00000926-D01P
10581-542EA 376 USD
10581-542
Anti-CD8B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD8B
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR