Anti-CD7 Mouse Polyclonal Antibody

Supplier: Abnova
H00000924-B01P
10508-882EA 335 USD
10508-882
Anti-CD7 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD7
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR