Anti-CD6 Mouse Polyclonal Antibody

H00000923-B01P
10560-202EA 316.92 USD
10560-202
Anti-CD6 Mouse Polyclonal Antibody
Type: Primary
Antigen: CD6
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR