Anti-ARR3 Mouse Monoclonal Antibody [clone: 2D7]

Supplier: Abnova
H00000407-M07
10581-000EA 335 USD
10581-000
Anti-ARR3 Mouse Monoclonal Antibody [clone: 2D7]
Antibodies
Type: Primary
Antigen: ARR3
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR