Anti-PHOX2A Mouse Monoclonal Antibody [clone: 4F6]

Supplier: Abnova
H00000401-M01
10596-898EA 335 USD
10596-898
Anti-PHOX2A Mouse Monoclonal Antibody [clone: 4F6]
Antibodies
Type: Primary
Antigen: PHOX2A
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR