Anti-ARHGDIG Mouse Monoclonal Antibody [clone: 1D11]

Supplier: Abnova
H00000398-M01
10577-226EA 335 USD
10577-226
Anti-ARHGDIG Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
Type: Primary
Antigen: ARHGDIG (Rho GDP dissociation inhibitor (GDI) gamma)
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR