Anti-ARHGDIA Mouse Polyclonal Antibody

H00000396-B02P
10509-408EA 316.92 USD
10509-408
Anti-ARHGDIA Mouse Polyclonal Antibody
Type: Primary
Antigen: ARHGDIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR