Anti-RHOA Mouse Monoclonal Antibody [clone: 1C3]

Supplier: Abnova
H00000387-M08
10599-718EA 335 USD
10599-718
Anti-RHOA Mouse Monoclonal Antibody [clone: 1C3]
Antibodies
Type: Primary
Antigen: RHOA
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR