Anti-ARG2 Mouse Monoclonal Antibody [clone: 3D10]

Supplier: Abnova
H00000384-M05
10580-978EA 335 USD
10580-978
Anti-ARG2 Mouse Monoclonal Antibody [clone: 3D10]
Antibodies
Type: Primary
Antigen: ARG2
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR