Anti-ARF5 Mouse Monoclonal Antibody [clone: 1B6]

Supplier: Abnova
H00000381-M02
10577-194EA 335 USD
10577-194
Anti-ARF5 Mouse Monoclonal Antibody [clone: 1B6]
Antibodies
Type: Primary
Antigen: ARF5
Clonality: Monoclonal
Clone: 1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR