Anti-AREG Mouse Monoclonal Antibody [clone: 3E4]

Supplier: Abnova
H00000374-M06
10577-188EA 335 USD
10577-188
Anti-AREG Mouse Monoclonal Antibody [clone: 3E4]
Antibodies
Type: Primary
Antigen: AREG
Clonality: Monoclonal
Clone: 3E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR