Anti-TRIM23 Mouse Monoclonal Antibody [clone: 2H4]

Supplier: Abnova
H00000373-M01
10604-628EA 335 USD
10604-628
Anti-TRIM23 Mouse Monoclonal Antibody [clone: 2H4]
Antibodies
Type: Primary
Antigen: TRIM23
Clonality: Monoclonal
Clone: 2H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR