Anti-ARAF Mouse Monoclonal Antibody [clone: 6H6]

Supplier: Abnova
H00000369-M01
10577-174EA 335 USD
10577-174
Anti-ARAF Mouse Monoclonal Antibody [clone: 6H6]
Antibodies
Type: Primary
Antigen: ARAF
Clonality: Monoclonal
Clone: 6H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR