Anti-ABCC6 Mouse Polyclonal Antibody

H00000368-B01P
10510-018EA 316.92 USD
10510-018
Anti-ABCC6 Mouse Polyclonal Antibody
Type: Primary
Antigen: ABCC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR