Anti-AR Mouse Monoclonal Antibody [clone: 1G3]

Supplier: Abnova
H00000367-M01
10576-884EA 335 USD
10576-884
Anti-AR Mouse Monoclonal Antibody [clone: 1G3]
Antibodies
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR