Anti-FASLG Mouse Monoclonal Antibody [clone: 3F3]

Supplier: Abnova
H00000356-M03
10604-298EA 335 USD
10604-298
Anti-FASLG Mouse Monoclonal Antibody [clone: 3F3]
Antibodies
Type: Primary
Antigen: FASLG (Fas ligand (TNF superfamily, member 6))
Clonality: Monoclonal
Clone: 3F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR