Anti-FAS Mouse Monoclonal Antibody [clone: 5B4]

Supplier: Abnova
H00000355-M01
10560-636EA 335 USD
10560-636
Anti-FAS Mouse Monoclonal Antibody [clone: 5B4]
Antibodies
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: Monoclonal
Clone: 5B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR