Anti-FAS Mouse Polyclonal Antibody

Supplier: Abnova
H00000355-A01
10560-638EA 243 USD
10560-638
Anti-FAS Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR