Anti-APP Mouse Monoclonal Antibody [clone: 2C12]

Supplier: Abnova
H00000351-M01
10577-164EA 335 USD
10577-164
Anti-APP Mouse Monoclonal Antibody [clone: 2C12]
Antibodies
Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR