Anti-APOA2 Mouse Monoclonal Antibody [clone: 1H6]

Supplier: Abnova
H00000336-M03
10577-098EA 335 USD
10577-098
Anti-APOA2 Mouse Monoclonal Antibody [clone: 1H6]
Antibodies
Type: Primary
Antigen: APOA2
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR