Anti-XIAP Mouse Monoclonal Antibody [clone: 2B3]

Supplier: Abnova
H00000331-M01
10551-466EA 335 USD
10551-466
Anti-XIAP Mouse Monoclonal Antibody [clone: 2B3]
Antibodies
Type: Primary
Antigen: XIAP
Clonality: Monoclonal
Clone: 2B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR