![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | BIRC3 |
Antigen Name | Baculoviral IAP Repeat Containing 3 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
Antigen Synonyms | HIAP1, CIAP2, RNF49, HAIP1, MALT2, AIP1, API2, MIHC, c-IAP2 |
Gene ID | 330 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |