Anti-APEX1 Mouse Monoclonal Antibody [clone: 3E12]

Supplier: Abnova
H00000328-M05
10577-046EA 335 USD
10577-046
Anti-APEX1 Mouse Monoclonal Antibody [clone: 3E12]
Antibodies
Type: Primary
Antigen: APEX1
Clonality: Monoclonal
Clone: 3E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR