Anti-APCS Mouse Monoclonal Antibody [clone: 4F3]

Supplier: Abnova
H00000325-M11
10577-040EA 335 USD
10577-040
Anti-APCS Mouse Monoclonal Antibody [clone: 4F3]
Antibodies
Type: Primary
Antigen: APCS
Clonality: Monoclonal
Clone: 4F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR