Anti-APBB2 Mouse Monoclonal Antibody [clone: 2D8]

Supplier: Abnova
H00000323-M01
10576-856EA 335 USD
10576-856
Anti-APBB2 Mouse Monoclonal Antibody [clone: 2D8]
Antibodies
Type: Primary
Antigen: APBB2
Clonality: Monoclonal
Clone: 2D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR