Anti-AOC2 Mouse Polyclonal Antibody

H00000314-B01
10559-006EA 310.77 USD
10559-006
Anti-AOC2 Mouse Polyclonal Antibody
Type: Primary
Antigen: AOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR