![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ANXA7 |
Antigen Name | Annexin A7 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | SNX, ANX7, Annexin-7, Annexin A7, Annexin VII, Synexin |
Gene ID | 310 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |