Anti-ANXA5 Mouse Monoclonal Antibody [clone: 1F4-1A5]

Supplier: Abnova
H00000308-M01
10577-010EA 335 USD
10577-010
Anti-ANXA5 Mouse Monoclonal Antibody [clone: 1F4-1A5]
Antibodies
Type: Primary
Antigen: ANXA5
Clonality: Monoclonal
Clone: 1F4-1A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR