![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ANXA5 |
Antigen Name | Annexin A5 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | ENX2, PP4, ANX5, RPRGL3, HEL-S-7 |
Gene ID | 308 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |