Anti-ANXA4 Mouse Monoclonal Antibody [clone: 1D3]

Supplier: Abnova
H00000307-M13
10577-004EA 335 USD
10577-004
Anti-ANXA4 Mouse Monoclonal Antibody [clone: 1D3]
Antibodies
Type: Primary
Antigen: ANXA4
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR