Anti-ANXA2 Mouse Monoclonal Antibody [clone: 3E8-B6]

Supplier: Abnova
H00000302-M01
10576-992EA 335 USD
10576-992
Anti-ANXA2 Mouse Monoclonal Antibody [clone: 3E8-B6]
Antibodies
Type: Primary
Antigen: ANXA2
Clonality: Monoclonal
Clone: 3E8-B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR