![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ANXA2 |
Antigen Name | Annexin A2 |
Reactivity | Rat, Mouse, Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
ELISA | Yes |
Antigen Synonyms | p36, ANXA2, CAL1H, ANX2L4, Annexin II, ANX2, Chromobindin-8, Lipocortin II, LPC2D, Annexin-2, Placental anticoagulant protein IV, PAP-IV, Protein I, Calpactin I heavy chain |
Gene ID | 302 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |