Anti-SLC25A6 Rabbit Polyclonal Antibody

Supplier: Abnova
H00000293-D01P
10601-030EA 376 USD
10601-030
Anti-SLC25A6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC25A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR