Anti-ANK1 Mouse Monoclonal Antibody [clone: 3C3]

Supplier: Abnova
H00000286-M01
10576-938EA 335 USD
10576-938
Anti-ANK1 Mouse Monoclonal Antibody [clone: 3C3]
Antibodies
Type: Primary
Antigen: ANK1
Clonality: Monoclonal
Clone: 3C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR