![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | BIN1 |
Antigen Name | Bridging integrator 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | 2C7 |
Host | Mouse |
Western Blot | Yes |
ELISA | Yes |
Antigen Synonyms | SH3P9, box-dependent myc-interacting protein 1, AMPHL, box dependant MYC interacting protein 1, amphiphysin-like protein, myc box-dependent-interacting protein 1, amphiphysin II, AMPH2 |
Gene ID | 274 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |