Anti-BIN1 Mouse Monoclonal Antibody [clone: 1H1]

Supplier: Abnova
H00000274-M01
10581-154EA 335 USD
10581-154
Anti-BIN1 Mouse Monoclonal Antibody [clone: 1H1]
Antibodies
Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 1H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR