Anti-AMPH Mouse Monoclonal Antibody [clone: 1H3-1D11]

Supplier: Abnova
H00000273-M01A
10623-844EA 335 USD
10623-844
Anti-AMPH Mouse Monoclonal Antibody [clone: 1H3-1D11]
Antibodies
Type: Primary
Antigen: AMPH
Clonality: Monoclonal
Clone: 1H3-1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR