![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | AMELX |
Antigen Name | Amelogenin X-linked |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Antigen Synonyms | AMG, AMGX, ALGN, AI1E, AIH1, AMGL |
Gene ID | 265 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |