Anti-AMBP Mouse Monoclonal Antibody [clone: 2E8]

Supplier: Abnova
H00000259-M08
10575-870EA 335 USD
10575-870
Anti-AMBP Mouse Monoclonal Antibody [clone: 2E8]
Antibodies
Type: Primary
Antigen: AMBP
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR