Anti-ALOX15 Mouse Monoclonal Antibody [clone: 3D8]

Supplier: Abnova
H00000246-M04
10580-910EA 335 USD
10580-910
Anti-ALOX15 Mouse Monoclonal Antibody [clone: 3D8]
Antibodies
Type: Primary
Antigen: ALOX15
Clonality: Monoclonal
Clone: 3D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR