Anti-ALOX12 Mouse Polyclonal Antibody

H00000239-B01P
10510-764EA 316.92 USD
10510-764
Anti-ALOX12 Mouse Polyclonal Antibody
Type: Primary
Antigen: ALOX12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR