Anti-ALDOB Mouse Monoclonal Antibody [clone: 3B3]

Supplier: Abnova
H00000229-M01
10576-696EA 335 USD
10576-696
Anti-ALDOB Mouse Monoclonal Antibody [clone: 3B3]
Antibodies
Type: Primary
Antigen: ALDOB
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR