![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ALDOA |
Antigen Name | Aldolase A |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | 3C12-6D11 |
Host | Mouse |
Western Blot | Yes |
ELISA | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | ALDA, HEL-S-87p, GSD12 |
Gene ID | 226 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |