Anti-ALDH3B2 Mouse Monoclonal Antibody [clone: 3E6]

Supplier: Abnova
H00000222-M01
10580-908EA 335 USD
10580-908
Anti-ALDH3B2 Mouse Monoclonal Antibody [clone: 3E6]
Antibodies
Type: Primary
Antigen: ALDH3B2
Clonality: Monoclonal
Clone: 3E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR