Anti-ALDH1B1 Mouse Monoclonal Antibody [clone: 2C7]

Supplier: Abnova
H00000219-M02
10576-658EA 335 USD
10576-658
Anti-ALDH1B1 Mouse Monoclonal Antibody [clone: 2C7]
Antibodies
Type: Primary
Antigen: ALDH1B1
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR