Anti-ALDH2 Mouse Monoclonal Antibody [clone: 4C11]

Supplier: Abnova
H00000217-M17
10576-666EA 335 USD
10576-666
Anti-ALDH2 Mouse Monoclonal Antibody [clone: 4C11]
Antibodies
Type: Primary
Antigen: ALDH2
Clonality: Monoclonal
Clone: 4C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR