Anti-ALDH2 Mouse Monoclonal Antibody [clone: 1E5]

Supplier: Abnova
H00000217-M01
10576-660EA 335 USD
10576-660
Anti-ALDH2 Mouse Monoclonal Antibody [clone: 1E5]
Antibodies
Type: Primary
Antigen: ALDH2
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR