Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: 1G6]

Supplier: Abnova
H00000216-M05
10576-652EA 335 USD
10576-652
Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: 1G6]
Antibodies
Type: Primary
Antigen: ALDH1A1
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR