Anti-ABCD1 Mouse Monoclonal Antibody [clone: 4B5]

Supplier: Abnova
H00000215-M01
10575-988EA 335 USD
10575-988
Anti-ABCD1 Mouse Monoclonal Antibody [clone: 4B5]
Antibodies
Type: Primary
Antigen: ABCD1
Clonality: Monoclonal
Clone: 4B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR